Learn More
Bis(2,4-pentanedionato)nickel(II), TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Bis(2,4-pentanedionato)nickel(II) |
| CAS | 120156-44-7 |
| Molecular Formula | 2 C5 H8 O2 . H2 O . Ni |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/2C5H8O2.Ni.H2O/c2*1-4(6)3-5(2)7;;/h2*3,6H,1-2H3;;1H2/b2*4-3-;; |
| SMILES | O.[Ni].C\C(=C\C(=O)C)\O.C\C(=C\C(=O)C)\O |
| IUPAC Name | (Z)-4-hydroxypent-3-en-2-one;nickel;hydrate |
| Molecular Weight (g/mol) | 276.94 |
| Formula Weight | 276.051 |