missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AcroBiosystems Biotinylated Human CD209 / DC-SIGN Protein, AvitagTM,His Tag
Description
CD209 is also known as CLEC4L, DC-SIGN and CD209 antigen. Pathogen-recognition receptor expressed on the surface of immature dendritic cells (DCs) and involved in initiation of primary immune response. On DCs it is a high affinity receptor for ICAM2 and ICAM3 by binding to mannose-like carbohydrates. May act as a DC rolling receptor that mediates transendothelial migration of DC presursors from blood to tissues by binding endothelial ICAM2. Seems to regulate DC-induced T-cell proliferation by binding to ICAM3 on T-cells in the immunological synapse formed between DC and T-cells.
Specifications
Specifications
| Accession Number | Uniprot |
| Characterization | CD209 Structure This protein carries an Avi tag (AvitagTM) at the N-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 43.0 kDa. The protein migrates as 47-50 kDa when calibrated against under reducing (R) condition (SDS-PAGE) due to glycosylation. |
| Format | Powder |
| Formulation | Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4 with trehalose as protectant. Contact us for customized product form or formulation. |
| Quality Control Testing | Less than 1.0 EU per μg by the LAL method / rFC method. |
| Quantity | 25 μg |
| Source | Biotinylated Human CD209 Protein, Avitag,His Tag (CD9-H82Q3) is expressed from human 293 cells (HEK293). It contains AA Gln 59 - Ala 404 (Accession # ). Predicted N-terminus: Gly Request for sequence |
| Biological Activity | Immobilized SARS-CoV-2 Spike S1, His Tag (Cat. No. S1N-C52Ht) at 1 μg/mL (100 μL/well) can bind Biotinylated Human CD209 Protein, Avitag,His Tag (Cat. No. CD9-H82Q3) with a linear range of 0.07-0.313 μg/mL (QC tested). |
| Species | Human |
| Content And Storage | -20°C to -70°C for 12 months in lyophilized state; -70°C for 3 months under sterile conditions after reconstitution. For long term storage, the product should be stored at lyophilized state at -20°C or lower. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?