missing translation for 'onlineSavingsMsg'
Learn More

Biotin-conjugated KRT80 Rabbit Polyclonal Antibody, Proteintech

Product Code. 30237579 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30237579 20 μL 20µL
30234808 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30237579 Supplier Proteintech Supplier No. BIOTIN1683520UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The KRT80 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KRT80. This antibody recognizes human, mouse, rat antigen. The KRT80 antibody has been validated for the following applications: IHC analysis. This antibody is biotin.

Specifications

Antigen KRT80
Applications Immunohistochemistry
Classification Polyclonal
Concentration 1000 μg/mL
Conjugate Biotin
Gene Symbols KB20
Host Species Rabbit
Immunogen Recombinant Protein
Purification Method Antigen affinity purification
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144501
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Antibody Molecular Weight 47 kDa, 50 kDa
Product Type Conjugated Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.