Learn More
Sartorius™ Biosart™ Monitor
Product Code. 11823292 Shop All Sartorius Products
Change view
Click to view available options
Color:
Gray/White
Green/Dark Green
White/Black
Packaging:
2 layers of 24
4 bags of 12
48/Pk.
Unit Size:
Pack of 48
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Packaging unitSize
11823292 Green/Dark Green 4 bags of 12 Pack of 48
11832691 Gray/White 4 bags of 12 Pack of 48
10679792 White/Black 48/Pk. Pack of 48
11833292 Gray/White 2 layers of 24 Pack of 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11823292 Supplier Sartorius™ Supplier No. 164024706K
Microbiological and analytical filtration units offering superior performance
The system enables the user to select the optimal hardware and consumables for their needs in quality assurance. At the heart of the system is a stainless steel manifold designed to accommodate all types of filter holder and funnels. Please note: grey filters turn black when wet. Multi-branch system for routine colony or particle counting.
TRUSTED_SUSTAINABILITY
Specifications
Color Green/Dark Green
Sterility Sterile
No. per Case 48Pack
No. per Pack 48Pack
Packaging 4 bags of 12
Quantity 48Pack
Type Monitor
compliance-icons compliance-icons
Product Identifier
  • X48 BIOSART 100 0,45µM 47MM GREEN
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H228-Flammable solid.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: nitrocellulose