Learn More
Labnet™ BioFree™ Pipet Tips
Product Code. p-7224808
Click to view available options
Volume (Metric) Dispensing:
200 μL
1000 μL
Unit Size:
Each
Product Code. 11492027
Brand: Labnet™ P1000200
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Labnet™ BioFree™ Pipet Tips are designed to fit a wide variety of single- and multi-channel pipettors.
  • Labnet BioFree™ Pipet Tips are designed to fit a wide variety of single- and multi-channel pipettors. Flexible walls and a series of internal sealing rings ensure a secure fit and minimal forces to eject pipet tips.
    • Exceptional accuracy and precision
    • Univeral fit RNase, DNase and Pyrogen safe
    • Perfect for Labnet pipettors
  • High Quality, FDA Compliant Pipet Tips
    • Labnet BioFree™ Pipet Tips are manufactured from high quality, FDA compliant polypropylene in a clean room environment. Diamond polished, stainless steel molds produce a tip with exceptionally smooth surfaces and extreme accuracy. During the manufacturing process, each lot of tips is tested for accuracy and precision. Tips are also carefully inspected for uniformity and manufacturing irregularities.
    • Certified RNase, DNase, and Pyrogen safe.
  • To ensure successful steam sterilization of your pipet tips, please adhere to the following guidelines:
    • Autoclave all pipet tips at 121°C for 15 minutes, 15 psi/1 atm
    • Do not use a dry cycle, as this may cause deformation or weakening of the plastic
    • Allow temperature within the autoclave to return to at least 80°C before removing the product
    • Excessive exposure to autoclaving heat may cause warping or decreased structural integrity.
Specifications
Volume (Metric) 200 μL
Volume (Metric) Dispensing 200 μL
Autoclavable Autoclavable
Quantity 1000 Tips/Unit, 20 Units/Cs.
Product Type Pipet Tips
For Use With (Equipment) Universal Fit
Product Line BioFree
Material Polypropylene