Learn More
Corning™ BioCoat™ Angiogenesis System for Endothelial Cell Invasion
Description
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 Assay System, PET Membrane Contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat Fibrillar Collagen 24-well insert system
Specifications
Specifications
| Pore Size | 3 μm |
| Type | Membrane |
| For Use With (Application) | Endothelial Cell Invasion |
| Quantity | 1/Pk. |
| No. of Wells | 24 |