missing translation for 'onlineSavingsMsg'
Learn More

BI-D1870, MedChemExpress

Product Code. p-200098067
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16480367

Brand: Medchem Express HY10510

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BI-D1870 is an ATP-competitive, cell permeable and brain penetrated inhibitor of RSK isoforms, with IC50s of 31 nM/24 nM/18 nM/15 nM for RSK1/RSK2/RSK3/RSK4, respectively.

BI-D1870 is an ATP-competitive, cell permeable and brain penetrated inhibitor of RSK isoforms, with IC50s of 31 nM/24 nM/18 nM/15 nM for RSK1/RSK2/RSK3/RSK4, respectively.

Specifications

Chemical Name or Material BI-D1870
CAS 501437-28-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H23F2N5O2
Quantity 50 mg
Solubility Information DMSO : 5 mg/mL (12.77 mM; Need ultrasonic)
SMILES OC1=C(F)C=C(NC2=NC(N(CCC(C)C)C(C)C(N3C)=O)=C3C=N2)C=C1F
Molecular Weight (g/mol) 391.42
Formula Weight 391.42
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.