missing translation for 'onlineSavingsMsg'
Learn More

BI-4464, MedChemExpress

Product Code. 16495178
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495178 10 mg 10mg
16465876 100 mg 100mg
16447907 5 mg 5mg
16411407 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16495178 Supplier Medchem Express Supplier No. HY124625

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BI-4464 is a highly selective ATP competitive inhibitor of PTK2/FAK, with an IC50 of 17 nM. A PTK2 ligand for PROTAC.

BI-4464 is a highly selective ATP competitive inhibitor of PTK2/FAK, with an IC50 of 17 nM. A PTK2 ligand for PROTAC.

Specifications

Chemical Name or Material BI-4464
CAS 1227948-02-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C28H28F3N5O4
Quantity 10 mg
Solubility Information DMSO : 16.67 mg/mL (30.01 mM; Need ultrasonic)
SMILES O=C(NC1CCN(C)CC1)C2=CC=C(NC3=NC=C(C(F)(F)F)C(OC4=CC=CC(CC5)=C4C5=O)=N3)C(OC)=C2
Molecular Weight (g/mol) 555.55
Formula Weight 555.55
Percent Purity 99.27%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.