Learn More
beta-Glycerol Phosphate Disodium Salt Pentahydrate, TRC
Product Code. 30009650
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009650 50 g 50g
30135464 100 g 100g
30008261 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30009650 Supplier Toronto Research Chemicals Supplier No. TRCG59845050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13408-09-8
Molecular Formula C3 H7 O6 P . 2 Na
Molecular Weight (g/mol) 216.04
Synonym 1,2,3-Propanetriol, 2-(dihydrogen phosphate), sodium salt, hydrate (1:2:5) (ACI), 1,2,3-Propanetriol, 2-(dihydrogen phosphate), disodium salt, pentahydrate (9CI), Glycerol, 2-(dihydrogen phosphate), disodium salt, pentahydrate (8CI), Sodium β-glycerophosphate pentahydrate, Disodium 2-glycerophosphate pentahydrate, Disodium β-glycerin phosphate pentahydrate, Disodium β-glycerophosphate pentahydrate, Glycerol-2-phosphate disodium salt pentahydrate, β-Glycerophosphate disodium salt pentahydrate
IUPAC Name disodium;1,3-dihydroxypropan-2-yl phosphate
SMILES [Na+].[Na+].OCC(CO)OP(=O)([O-])[O-]
Specifications
Quantity 50 g
InChI Formula InChI=1S/C3H9O6P.2Na/c4-1-3(2-5)9-10(6,7)8;;/h3-5H,1-2H2,(H2,6,7,8);;/q;2*+1/p-2
Formula Weight 215.9776
Chemical Name or Material beta-Glycerol Phosphate Disodium Salt Pentahydrate