missing translation for 'onlineSavingsMsg'
Learn More

Bestatin Free Acid MP Biomedicals

Product Code. 11404001 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 11404001

Brand: MP Biomedicals 0215284405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 58970-76-6
Molecular Formula C16H24N2O4
MDL Number MFCD00083262
Quantity 5 mg
Synonym bestatin, ubenimex, ubenimexum latin, ubenimex inn:jan, bestatin;ubenimex, s-2-2s,3r-3-amino-2-hydroxy-4-phenylbutanamido-4-methylpentanoic acid, ubenimex bestatin, chembl29292, 2s,3r-3-amino-2-hydroxy-4-phenylbutanoyl-l-leucine, 3-r-amino-2-s-hydroxy-4-phenylbutanoyl-s-leucine
InChI Key VGGGPCQERPFHOB-MCIONIFRSA-N
SMILES CC(C)C[C@@H](NC(=O)[C@@H](O)[C@H](N)CC1=CC=CC=C1)C(O)=O
IUPAC Name (2R)-2-[(2S,3R)-3-amino-2-hydroxy-4-phenylbutanamido]-4-methylpentanoic acid
Molecular Weight (g/mol) 308.38
PubChem CID 72172
Formula Weight 308.4
Chemical Name or Material Bestatin, Free Acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.