missing translation for 'onlineSavingsMsg'
Learn More

Berberine Sulfate, TRC

Product Code. 30337297
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30337297 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30337297 Supplier Toronto Research Chemicals Supplier No. TRCB3181551G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Berberine Sulfate
CAS 633-66-9
Molecular Formula C20 H18 N O4 . H S O4
Quantity 1 g
Synonym 7,8,13,13a-tetradehydro-9,10-dimethoxy-2,3-(methylenedioxy)Berbinium Sulfate, Berberine Bisulfate, Berberine Hemisulfate, Berberine Hydrogen Sulfate, Berberine Sulfate, Berberine Sulfate (1:1), NSC 150444
InChI Formula InChI=1S/C20H18NO4.H2O4S/c1-22-17-4-3-12-7-16-14-9-19-18(24-11-25-19)8-13(14)5-6-21(16)10-15(12)20(17)23-2;1-5(2,3)4/h3-4,7-10H,5-6,11H2,1-2H3;(H2,1,2,3,4)/q+1;/p-1
SMILES COc1ccc2cc3c4cc5OCOc5cc4CC[n+]3cc2c1OC.OS(=O)(=O)[O-]
IUPAC Name 16,17-dimethoxy-5,7-dioxa-13-azoniapentacyclo[11.8.0.02,10.04,8.015,20]henicosa-1(13),2,4(8),9,14,16,18,20-octaene;hydrogen sulfate
Molecular Weight (g/mol) 433.432
Formula Weight 433.083
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.