missing translation for 'onlineSavingsMsg'
Learn More

Benzyl cinnamate, 99%

Product Code. p-7024601
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11464117

Brand: Thermo Scientific Alfa Aesar A19550.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-41-3 | C16H14O2 | 238.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-41-3
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.29
MDL Number MFCD00004789,MFCD00004789,MFCD00004789
InChI Key NGHOLYJTSCBCGC-VAWYXSNFSA-N
Synonym benzyl alcohol cinnamic ester, benzylcinnamate, benzyl alcohol, cinnamate, wln: r1u1vo1r, benzyl .gamma.-phenylacrylate, benzyl cis-cinnamate, benzyl z-cinnamate, 2-propenoic acid, 3-phenyl-, phenylmethyl ester, z-benzyl 3-phenylacrylate, cinnamic acid, benzyl ester, z
PubChem CID 15558051
IUPAC Name benzyl (Z)-3-phenylprop-2-enoate
SMILES O=C(OCC1=CC=CC=C1)\C=C\C1=CC=CC=C1

Specifications

Melting Point 34°C to 36°C
Density 1.11
Boiling Point 195°C to 200°C (5 mmHg)
Flash Point 180°C (356°F)
Quantity 100 g
Beilstein 2051339
Merck Index 14,1130
Formula Weight 238.29
Percent Purity 99%
Chemical Name or Material Benzyl cinnamate
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Skin sensitiser Category 1B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.