missing translation for 'onlineSavingsMsg'
Learn More

Benzotrichloride, 98%

Product Code. p-7022749
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
This item is not returnable. View return policy

Product Code. 11447567

Brand: Thermo Scientific Alfa Aesar A15652.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 98-07-7 | C7H5Cl3 | 195.47 g/mol

It finds its use in other preparations of substituted benzophenones, for benzoylation of phenols under basic, phase-transfer conditions. D,D,D-Trichlorotoluene is an intermediate, used exclusively in the industrial production of other intermediates such as Benzoylchloride, Benzotrifluoride and 2,4-Dihydroxybenzophenone. α,α,α-Trichlorotoluene is also used in determination of chlorinated toluenes in effluents of wastewater treatment plants and textile industries by closed loop stripping analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its use in other preparations of substituted benzophenones, for benzoylation of phenols under basic, phase-transfer conditions. D,D,D-Trichlorotoluene is an intermediate, used exclusively in the industrial production of other intermediates such as Benzoylchloride, Benzotrifluoride and 2,4-Dihydroxybenzophenone. α,α,α-Trichlorotoluene is also used in determination of chlorinated toluenes in effluents of wastewater treatment plants and textile industries by closed loop stripping analysis.

Solubility
Solubility in water is 0.1 g/L at 20°C.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-07-7
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.47
MDL Number MFCD00000786
InChI Key XEMRAKSQROQPBR-UHFFFAOYSA-N
Synonym benzotrichloride, trichloromethyl benzene, alpha,alpha,alpha-trichlorotoluene, phenylchloroform, toluene trichloride, benzenyl trichloride, benzene, trichloromethyl, benzyl trichloride, benzylidyne chloride, phenyltrichloromethane
PubChem CID 7367
ChEBI CHEBI:82274
IUPAC Name trichloromethylbenzene
SMILES ClC(Cl)(Cl)C1=CC=CC=C1

Specifications

Melting Point -8°C to -0.5°C
Density 1.38
Boiling Point 90°C to 91°C (15 mmHg)
Flash Point 97°C (207°F)
Refractive Index 1.557
Quantity 500 g
UN Number UN2226
Beilstein 508152
Sensitivity Moisture sensitive
Merck Index 14,1109
Formula Weight 195.48
Percent Purity 98%
Chemical Name or Material Benzotrichloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.