missing translation for 'onlineSavingsMsg'
Learn More

Benzhydrol, 99%

Product Code. p-7020808
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11495323

Brand: Thermo Scientific Alfa Aesar A12884.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91-01-0 | C13H12O | 184.238 g/mol

Benzhydrol is widely used as intermediates in pharmaceuticals (including antihistamines), agrochemicals, perfumes and other organic compounds. It is used as a fixative in the perfume industry. It is involved in polymerization reaction as a terminating group. It is used as precursor to prepare modafinil, benztropine and diphehydramine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzhydrol is widely used as intermediates in pharmaceuticals (including antihistamines), agrochemicals, perfumes and other organic compounds. It is used as a fixative in the perfume industry. It is involved in polymerization reaction as a terminating group. It is used as precursor to prepare modafinil, benztropine and diphehydramine.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents, acids, acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-01-0
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.238
MDL Number MFCD00004488
InChI Key QILSFLSDHQAZET-UHFFFAOYSA-N
Synonym benzhydrol, diphenylcarbinol, benzohydrol, benzhydryl alcohol, hydroxydiphenylmethane, diphenylmethyl alcohol, diphenyl carbinol, alpha-phenylbenzenemethanol, diphenyl-methanol, benzenemethanol, .alpha.-phenyl
PubChem CID 7037
IUPAC Name diphenylmethanol
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)O

Specifications

Melting Point 64°C to 68°C
Boiling Point 297°C to 298°C
Flash Point 160°C (320°F)
Odor Odorless
Quantity 50 g
Beilstein 1424379
Merck Index 14,1090
Solubility Information Slightly soluble in water.
Formula Weight 184.24
Percent Purity 99%
Chemical Name or Material Benzhydrol
Show More Show Less
compliance-icons
Product Identifier
  • Benzhydrol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Benzhydrol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.