Learn More
Lenz Laborglasintrumente™ Bent Connector
Brand:  Lenz Laborglasintrumente™ 05313044
Additional Details : Weight : 0.50000kg
Product Code. 10709411
  • 69.91 EUR / Each
Estimated Shipment: 26-06-2024
Description
  • With cone and lateral socket NS 29/32
  • With socket NS 14/23 (for thermometer or air leak tube)
  • Made of DURAN™ tubing
Specifications
Specifications
Duran Tubing
1
NS 29/32