missing translation for 'onlineSavingsMsg'
Learn More

Benserazide hydrochloride, MedChemExpress

Product Code. 16410918
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16410918

Brand: Medchem Express HYB0404A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Benserazide hydrochloride (Serazide) is commonly used in Parkinson's disease and is an inhibitor of peripheral aromatic L-amino acid decarboxylase (AADC).

Benserazide hydrochloride (Serazide) is commonly used in Parkinson's disease and is an inhibitor of peripheral aromatic L-amino acid decarboxylase (AADC).

Specifications

Chemical Name or Material Benserazide hydrochloride
CAS 14919-77-8
Color White
Purity Grade Notes Research
Molecular Formula C10H16ClN3O5
Quantity 5 g
Synonym Serazide Ro 4-4602
Solubility Information DMSO : 100 mg/mL (340.48 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (170.24 mM)
SMILES NC(CO)C(NNCC1=CC=C(O)C(O)=C1O)=O.Cl
Molecular Weight (g/mol) 293.7
Formula Weight 293.7
Percent Purity 97.4%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.