missing translation for 'onlineSavingsMsg'
Learn More

Benin, MedChemExpress

Product Code. p-200098560
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16492338

Brand: Medchem Express HYU00073

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Benin is a potent cytostatic drug that can be used for the treatment of generalized carcinoma of the breast.

Benin is a potent cytostatic drug that can be used for the treatment of generalized carcinoma of the breast.

Specifications

Chemical Name or Material Benin
CAS 22181-94-8
Color White
Purity Grade Notes Research
Molecular Formula C14H19N5O3S
Quantity 5 mg
Synonym Butocin Butocine
SMILES O=C(OCC)CNC(CCCCSC1=C2N=CNC2=NC=N1)=O
Molecular Weight (g/mol) 337.4
Formula Weight 337.4
Percent Purity 97.85%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.