Learn More
WTW™ Benchtop Tripod Stand for WTW™ pH Electrodes
Click to view available options
For Use With (Equipment):
pH electrodes, ion-sensitive electrodes, reference electrodes, temperature sensors, oxygen sensors and TetraCon 325 conductivity cells
Unit Size:
Each
Specifications
Specifications
| Type | Benchtop Tripod Stand |
| For Use With (Equipment) | pH electrodes, ion-sensitive electrodes, reference electrodes, temperature sensors, oxygen sensors and TetraCon 325 conductivity cells |