Learn More
Benchmark Scientific™ Benchmark Industrial HCS High Capacity Stirrers
Click to view available options
Capacity:
50 L
150 L
Unit Size:
Each
Description
- Stir up to 50 or 150 L volumes
- Chemical resistant, white ceramic top surface
- Powerful motor ideal for large vessel applications
- Digital control of both stirring speed and timer
Specifications
Specifications
| Speed | 50 to 1200 rpm |
| Stirring Positions | 1 |
| Top Plate Material | Ceramic-coated Aluminum |
| Capacity | 150 L |
| Display | Digital |
| Shape | Square |
| Voltage | 230 V |
| Wattage | 80 W |
| Dimensions (L x W x H) | 62 x 50 x 16.5 cm |
| Plug Type | EU |
| Show More |