Learn More
Nabertherm™ Bench-Top Forced Convection Chamber Furnace
Shop All Nabertherm Products
Click to view available options
Capacity (Metric):
15 L
50 L
Controller Type:
B510
C550
Unit Size:
Each
Description
- Max temp 850°C
- Horizontal air circulation with optimum distribution through stainless steel baffles
- Swing door hinged on the right
Specifications
Specifications
| Controller Type | B510 |
| Capacity (Metric) | 50 L |
| Temperature (Metric) | 850°C |
| Type | Furnace |