Learn More
Belotecan hydrochloride, MedChemExpress
Product Code. 16476226
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16476226
Brand: Medchem Express HY13566A
MedChemExpress Belotecan hydrochloride (CKD-602 hydrochloride), a Topoisomerase I inhibitor, is a synthetic camptothecin derivative.
Belotecan hydrochloride (CKD-602 hydrochloride), a Topoisomerase I inhibitor, is a synthetic camptothecin derivative.
Specifications
Chemical Name or Material Belotecan hydrochloride
Physical Form Solid
CAS 213819-48-8
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C25H28ClN3O4
For Use With (Application) Cancer-programmed cell death
Synonym CKD-602
Solubility Information DMSO : 25 mg/mL (53.20 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C1[C@](O)(CC)C2=C(CO1)C(N3CC4=C(CCNC(C)C)C5=CC=CC=C5N=C4C3=C2)=O.[H]Cl
Molecular Weight (g/mol) 469.96
Formula Weight 469.96
Percent Purity 98.25%
Grade Research