missing translation for 'onlineSavingsMsg'
Learn More

Belnacasan, MedChemExpress

Product Code. 16471717
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16471717

Brand: Medchem Express HY13205

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Belnacasan (VX-765) is an orally bioactive prodrug of VRT-043198, which is a potent and selective inhibitor of IL-converting enzyme (ICE)/caspase-1 with Kis of 0.8 nM and less than 0.6 nM for caspase-1 and caspase-4, respectively. Belnacasan (VX-765) inhibits the release of LPS-induced IL-1β and IL-18 by human PBMCs with an IC50 of ∼0.7 μM.

Belnacasan (VX-765) is an orally bioactive prodrug of VRT-043198, which is a potent and selective inhibitor of IL-converting enzyme (ICE)/caspase-1 with Kis of 0.8 nM and less than 0.6 nM for caspase-1 and caspase-4, respectively. Belnacasan (VX-765) inhibits the release of LPS-induced IL-1β and IL-18 by human PBMCs with an IC50 of ∼0.7 μM.

Specifications

Chemical Name or Material Belnacasan
Physical Form Powder
CAS 273404-37-8
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C24H33ClN4O6
For Use With (Application) COVID-19-immunoregulation
Synonym VX-765
Solubility Information DMSO : 260 mg/mL (510.81 mM; Need ultrasonic) ∣H2O : 1 mg/mL (1.96 mM; ultrasonic and warming and heat to 60°C)
SMILES ClC1=C(N)C=CC(C(N[C@@H](C(C)(C)C)C(N2[C@H](C(N[C@H]3CC(O[C@H]3OCC)=O)=O)CCC2)=O)=O)=C1
Molecular Weight (g/mol) 509.0
Formula Weight 509.0
Percent Purity 98.03%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.