Learn More
Bel-Art™ Plastic Cover Slip
Product Code. 12937320
Change view
Click to view available options
Quantity:
1000/Pk.
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12937320 1000/Pk. Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12937320 Supplier Bel-Art™ Supplier No. H446900000
Add to basket
Add to basket
Flexible vinyl cover slips are packaged with interleaved tissue for protection.
  • Provides a refractive index similar to glass
  • Unbreakable, safer than glass
  • Used in biological and medical labs as well as educational applications
Specifications
Product Type Coverslip
Shape Square
Material Vinyl
Length (Metric) 22 mm
Width (Metric) 22 mm
Thickness (Metric) 15 mm
For Use With (Application) For separated with tissues to prevent scratches
Quantity 1000/Pk.