Learn More
Bel-Art™ SP Scienceware™ Microtube Cutter
Safely, accurately and easily sections microcentrifuge tubes and eliminates the hazards of using razor blades or knives
14.95 EUR - 38.40 EUR
Specifications
For Use With Microtube cutter
View More Specs
Products 2
Product Code Brand Material Product Type Price Quantity & Availability  
Product Code Brand Material Product Type Price Quantity & Availability  
12395759
View Documents Promotion Details
Bel-Art™
F21018-0001
Steel Microtube Cutter Replacement Blade
14.95 EUR
Each
View alternative products
Save Up to 
»
Estimated Shipment: 04-07-2024
to see stock.
Add to basket
11730635
View Documents Promotion Details
Bel-Art™
F21018-0000
Chrome-plated Steel Microtube Cutter This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • Makes it easy to retrieve radioactive pellets from the tips of tubes without resuspension or the risk of splashed radioactive substances
  • Works well with polyethylene c or polypropylene microcentrifuge tubes up to 1.5mL
  • Chrome-plated body with replaceable steel blade
  • Maximum cut diameter is 9.4mm (0.370 in.)
Not recommended for polystyrene tubes.
Specifications
Specifications
Microtube cutter