Learn More
Bel-Art™ SP Scienceware™ Fritware™ Porous Polyethylene Sheets
Product Code. 11392224 Shop All SP Scienceware Products
Click to view available options
Porosity:
Coarse
Medium
Dimensions (L x W):
18 x 18 in. (46 x 46cm)
Thickness (Metric):
1.6 mm
3.2 mm
Unit Size:
Each
Product Code. 11392224
Brand: Bel-Art™ H136380518
Supplied in medium or coarse porosity
  • Can be cut, sawed, formed, machined and welded to fabricate special items made partly or wholly of this porous material
  • Medium porosity style has an average pore size of 70 microns
  • Coarse porosity style averages 110 microns
Specifications
Filter Type Microporous Filter
Format Sheet
Material HDPE
Material (Membrane) Polyethylene
Pore Size 70 μm
Sterility Non-sterile
For Use With (Application) Can be cut, sawed, formed, machined and welded to fabricate special items made partly or wholly of this porous material
Porosity Medium
Thickness (Metric) 3.2 mm
Type Porous Polyethylene Sheet
Dimensions (L x W) 18 x 18 in. (46 x 46cm)
Length (English) 18 in.
Length (Metric) 46 cm
Width (English) 18 in.
Width (Metric) 46 cm