Learn More
Bel-Art™ Dispensing Drop Bottles
Product Code. 12388218 Shop All SP Scienceware Products
Change view
Click to view available options
Capacity (Metric):
15 mL
30 mL
60 mL
125 mL
185 mL
250 mL
500 mL
Unit Size:
Pack of 12
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
12388218 250 mL Pack of 12
12338218 15 mL Pack of 12
12348218 30 mL Pack of 12
12358218 60 mL Pack of 12
12368218 125 mL Pack of 12
12378218 185 mL Pack of 12
12398218 500 mL Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12388218 Supplier Bel-Art™ Supplier No. F116370008
Only null left
Add to basket
Add to basket
Easy-squeeze dispensing bottles allow for critical drop control.
  • Sealer cap snaps on easily and continues to hold a tight seal even after repeated use
  • Tip opening is 0.76 mm (0.030 in.)
  • Molded from low density polyethylene (LDPE)
Specifications
Capacity 8 oz., 250 mL
Capacity (Metric) 250 mL
Capacity (English) 8 oz.
Color Translucent, Red
Material LDPE
No. per Pack 12
Sterility Non-sterile
Closure Included Yes
Disposable Reusable
Dropper Capacity 8 oz.
Drop Size 0.002 oz.
Graduated No
Mouth Narrow, Threaded
Shape Cylindrical
Height (Metric) 165 mm
Product Type Dispensing Bottle
Width (English) 2.3 in.
Length (English) 6.5 in.
Height (English) 6.5 in.
Closure Material Polyethylene
Closure Size 24 mm
Diameter (Metric) 58.4 mm
Closure Type Snap