Learn More
Invitrogen™ Custom Beacon Primer
Product Code. 10012282
Click to view available options
Quantity:
200 nmol
Unit Size:
Each
Product Code. 10012282
Brand: Invitrogen™ 11848017
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole.
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Quantity 200 nmol