missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BDP Recombinant Rabbit Monoclonal Antibody (008)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535969
This item is not returnable.
View return policy
Description
BDP Recombinant Monoclonal Antibody for Western Blot
This gene encodes a member of the ARID family of DNA-binding proteins. The encoded protein is homologous with two proteins that bind to the retinoblastoma gene product, and also with the mouse Bright and Drosophila dead ringer proteins. A pseudogene on chromosome 1p31 exists for this gene. Members of the ARID family have roles in embryonic patterning, cell lineage gene regulation, cell cycle control, transcriptional regulation and possibly in chromatin structure modification.
Specifications
| BDP | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q8IVW6 | |
| ARID3B | |
| A synthetic peptide corresponding to the center region of the Human ARID3B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| 008 | |
| Unconjugated | |
| ARID3B | |
| ARID domain-containing protein 3B; ARID3B; AT rich interactive domain 3B (Bright like); AT rich interactive domain 3B (BRIGHT- like); AT rich interactive domain 3B (BRIGHT-like); AT-rich interaction domain 3B; AT-rich interactive domain-containing protein 3B; Bdp; bright and dead ringer gene product homologous protein Bdp; bright and dead ringer protein; Bright-like; Bright-like protein; dead ringer homolog 2; Dri2; DRIL2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10620 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction