missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Mouse MCP-1 ELISA Set
Click to view available options
Quantity:
20 Plates
Unit Size:
1 set
Description
- Contains the components necessary to develop enzymelinked immunosorbent assays (ELISA) for natural or recombinant mouse MCP-1 in serum, plasma, and cell culture supernatants
Enzyme Linked Immunosorbent Assay
Specifications
Specifications
| Product Type | Mouse MCP-1 ELISA set |
| Sufficient For | 20 96-well Plates |
| For Use With (Application) | To develop ELISA for natural or recombinant Mouse MCP-1 in Serum, Plasma and cell culture supernatants |
| Kit Contents | anti-Mouse MCP-1 monoclonal (capture) antibody, anti-Mouse MCP-1 monoclonal (detection) antibody Conjugated to Horseradish Peroxidase, Instruction/analysis certificate, Lyophilized recombinant Mouse MCP-1 (standard) |
| Quantity | 20 Plates |
| Storage Requirements | 2°C to 8°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction