Learn More
BD Mouse IL-4 ELISA Set
Click to view available options
Quantity:
20 Plates
Unit Size:
1 set
Description
Includes
Capture antibody, detection antibody, enzyme reagent, standards, instruction/analysis certificate
To develop enzyme-linked immunosorbent assays (ELISA) for natural or recombinant mouse IL-4 in serum, plasma, and cell culture supernatants
Enzyme-linked immunosorbent assay
Specifications
Specifications
| Concentration | 7.8 to 500pg/mL |
| Product Type | Mouse IL-4 ELISA set |
| Sufficient For | 20 96-well Plates |
| For Use With (Application) | To develop ELISA for natural or recombinant Mouse IL-4 in Serum, Plasma and cell culture supernatants |
| Kit Contents | anti-Mouse IL-4 monoclonal (capture) antibody, Biotinylated anti-Mouse IL-4 monoclonal (detection) antibody, Instruction/analysis certificate, Lyophilized recombinant Mouse IL-4 (standard), Streptavidin-HRP Conjugate |
| Quantity | 20 Plates |
| Storage Requirements | 2°C to 8°C |