Learn More
BD Human IL-12 (p40) ELISA Set
Description
Includes
- Capture Antibody: Anti-human IL-12 monoclonal antibody
- Detection Antibody: Biotinylated anti-human IL-12 monoclonal antibody
- Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate (SAv-HRP)
- Standards: Recombinant human IL-12 (p40), lyophilized
Requires
96-well BD Falcon™ ELISA plates (Cat. No. 353279) are recommended, Microplate reader capable of measuring absorbance at 450nm, Precision pipettes, Graduated cylinder, one liter, Deionized or distilled water, Wash bottle or automated washer, Log-log graph paper or automated data reduction, Tubes to prepare standard dilutions, Laboratory timer, Plate sealers or parafilm
BD OptEIA™ Sets allow flexible assay design to fit individual laboratory needs. To design an immunoassay with different sensitivity and dynamic range
- Standard Range: 31.3 - 2000 pg/ml
- Calibrated against purified Baculovirus-expressed recombinant human IL-12 (p40)
ELISA
Specifications
Specifications
| Concentration | 31.3 to 2000pg/mL |
| Product Type | Human IL-12 (p40) ELISA set |
| Sufficient For | 20 96-well Plates |
| For Use With (Application) | To develop ELISA for natural or recombinant Human IL-12 (p40) in Serum, Plasma and cell culture supernatant |
| Kit Contents | anti-Human IL-12 monoclonal (capture) Antibody, Biotinylated anti-Human IL-12 monoclonal (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Human IL-12 (p40)(standard), Streptavidin-HRP Conjugate |
| Quantity | 20 Plates |
| Storage Requirements | 2°C to 8°C |
For research use only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.