Learn More
BD Difco™ HYcheck™ Contact Slides: Plate Count Agar with TTC
Click to view available options
:
Plate Count Agar with TTC; 10U
Description
- Designed for environmental testing
- Double sided
- Hinged paddle bends for easy sampling
Order Info
Ten units per pack.
Specifications
Specifications
| Color | Light Amber |
| Content And Storage | 2°C to 8°C |
| Form | Solid |
| Manufacturing Origin | United States |
| Media Type | Plate Count Agar |
| Formulation | Pancreatic digest of Casein 5g/L, Yeast extract 2.5g/L, Dextrose 1g/L, Agar 15g/L |
| For Use With (Application) | Used for assess the microbiological contamination of surfaces or fluids |
| Packaging Type | Slide |
| Quantity | 10 U |