Learn More
BD Difco™ Dehydrated Culture Media: Tryptose Broth
Click to view available options
:
Tryptose Broth; Quantity: 500g
Description
Cultivation of Brucella and a variety of pathogenic microorganisms
Certifications
BAM, CCAM, COMPF
Specifications
Specifications
| Certifications/Compliance | BAM, CCAM, COMPF |
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Tryptose Broth |
| Product Type | Microbiology Media |
| Formulation | Tryptose 20g/L, Dextrose 1g/L, Sodium Chloride 5g/L |
| For Use With (Application) | Cultivation of Brucella and a variety of pathogenic microorganisms |
| Packaging Type | Bottle |
| Quantity | 500 g |