missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: MSRV Medium

Product Code. 11788293
Click to view available options
:
MSRV Medium; 500g
This item is not returnable. View return policy

Product Code. 11788293

Brand: BD 218681

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For detecting motile Salmonella in feces and food products

  • Used with Novobiocin Antimicrobic Supplement

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CCAM
Content And Storage 2°C to 30°C
Form Dehydrated
Media Type MSRV Medium
Product Type MSRV Medium
Formulation Tryptose 4.59g/L, Casein Hydrolysate (acid) 4.59g/L, Sodium Chloride 7.34g/L, Monopotassium Phosphate 1.47g/L, Magnesium Chloride (anhydrous) 10.9g/L, Malachite green Oxalate 37mg/L, Agar 2.7g/L
For Use With (Application) Novobiocin antimicrobic supplement
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.