missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Micro Assay Culture Agar
Description
Order Info
See our web site or the Fisher Scientific™ HealthCare catalog for product information.
Specifications
Specifications
| Certifications/Compliance | CE |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | Micro Assay Culture Agar |
| Product Type | Agar |
| Formulation | Proteose Peptone 5g/L, Yeast extract 20g/L, Dextrose 10g/L, Monopotassium Phosphate 2g/L, Polysorbate 80 0.1g/L, Agar 10g/L |
| For Use With (Application) | Cultivation of lactoBacilli and other organisms used in microbiological assay |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?