Learn More
BD Difco™ Dehydrated Culture Media: Antibiotic Medium 11
Description
- Erythromycin/Neomycin Agar Microbiological assay of antibiotics
Specifications
Specifications
| Certifications/Compliance | AOAC, USDA, USP |
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | Antibiotic Medium 11 |
| Product Type | Antibiotic Medium |
| Formulation | Beef extract 1.5g/L, Yeast extract 3g/L, Pancreatic digest of Casein 4g/L, Peptone 6g/L, Dextrose 1g/L, Agar 15g/L |
| For Use With (Application) | Erythromycin/Neomycin Agar Microbiological assay of Antibiotics |
| Packaging Type | Bottle |
| Quantity | 500 g |