Learn More
Gibco™ Difco™ Tryptone Peptone
Product Code. 17787009
Change view
Click to view available options
Quantity:
1 lb.
5 lb.
25 lb.
Unit Size:
11.34kg
2.26kg
454g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17787009 5 lb. 2.26kg
16289761 1 lb. 454g
16209771 25 lb. 11.34kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17787009 Supplier Gibco™ Supplier No. 211922
Difco Tryptone Peptone is an animal origin (AO) pancreatic digest of casein. In addition to providing a unique nutritional profile, it has a higher nitrogen content than our other casein products. Tryptone Peptone supports the growth of many different types of bacteria and fungi, including fastidious organisms and in toxin production.
Difco Tryptone Peptone is an animal origin (AO) pancreatic digest of casein. In addition to providing a unique nutritional profile, it has a higher nitrogen content than our other casein products. Tryptone Peptone supports the growth of many different types of bacteria and fungi, including fastidious organisms and in toxin production. Customers have used this supplement in the development of animal health vaccines, such as Clostridial vaccines, and with human health Botox-like products produced from Clostridium botulinum.
Features:
  • Animal origin, pancreatic digest of casein
  • Primary nitrogen source in trypticase soy broth and agar
  • Referenced in Official Methods of Analysis of AOAC International
  • Supports the growth of many different types of bacteria and fungi
Applications of Tryptone Peptone include:
  • Recommended for use in media formulations when good growth of fungi and bacteria is required
  • Used in the development of animal health vaccines and with human health Botox-like products
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance LUO, MUO
Content And Storage Store at 2°C to 25°C in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Bacterial Media
Format Pail
For Use With (Application) Dehydrated culture media
Quantity 5 lb.
Source Animal Origin