Learn More
BD BBL™ Taxo™ Differentiation Discs, Novobiocin
Product Code. 11732994
Click to view available options
Concentration:
5 μg
Unit Size:
Pack of 50
Product Code. 11732994
Brand: BD 231750
For determination of novobiocin resistance in coagulase negative staphylococci
  • Absorbant paper containing 5μg novobiocin
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Novobiocin
Concentration 5 μg
Content And Storage -20°C to +8°C
Packaging Type Disc
Quantity 1 Pack
compliance-icons
Product Identifier
  • BD BBL Taxo Differentiation Discs Novobiocin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Novobiocin sodium
For in vitro Diagnostic Use.