missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Trimethoprim

Product Code. 17392381
Click to view available options
Concentration:
5 μg
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 17392381

Brand: BD 231601

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents

  • Clearly marked on both sides with letters and numbers designating agent and concentration

TRUSTED_SUSTAINABILITY

Specifications

Antimicrobial Agents Trimethoprim
Concentration 5 μg
Packaging Type Disc
Quantity 10/Pk.
compliance-icons
Product Identifier
  • Disc Trimethoprim Tmp-5 10 Ea
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Trimethoprim
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.