Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Tobramycin
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Description
- Clearly marked on both sides with letters and numbers designating agent and concentration
- Available in pack of 1 and 10
Specifications
Specifications
| Antimicrobial Agents | Tobramycin |
| Concentration | 10 μg |
| Packaging Type | Disc |
| Quantity | 10/Pk. |