Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Tobramycin
Product Code. 17352381
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Product Code. 17352381
Brand: BD 231569
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Clearly marked on both sides with letters and numbers designating agent and concentration
  • Available in pack of 1 and 10
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Tobramycin
Concentration 10 μg
Packaging Type Disc
Quantity 10/Pk.