missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Rifampin

Product Code. p-4897863
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 17332441

Brand: BD 231544

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents

  • Clearly marked on both sides with letters and numbers designating agent and concentration
  • Available in pack of 1 and 10

TRUSTED_SUSTAINABILITY

Specifications

Antimicrobial Agents Rifampin
Concentration 5 μg
Packaging Type Disc
Quantity 10/Pk.
compliance-icons
Product Identifier
  • Disc Rifampin Ra-5 10 Ea
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P233-Keep container tightly closed.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P262-Do not get in eyes, on skin, or on clothing.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P320-Specific treatment is urgent (see on this label).
  • P361-Take off immediately all contaminated clothing.
  • P363-Wash contaminated clothing before reuse.
  • P403-Store in a well-ventilated place.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : rifampicin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.