Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Rifampin
Product Code. 17332441
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Product Code. 17332441
Brand: BD 231544
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Clearly marked on both sides with letters and numbers designating agent and concentration
  • Available in pack of 1 and 10
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Rifampin
Concentration 5 μg
Packaging Type Disc
Quantity 10/Pk.
compliance-icons
Product Identifier
  • Disc Rifampin Ra-5 10 Ea
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P233-Keep container tightly closed.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P262-Do not get in eyes, on skin, or on clothing.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P320-Specific treatment is urgent (see on this label).
  • P361-Take off immediately all contaminated clothing.
  • P363-Wash contaminated clothing before reuse.
  • P403-Store in a well-ventilated place.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : rifampicin