Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Oxytetracycline
Product Code. 11702964
Click to view available options
Concentration:
30 μg
Unit Size:
Pack of 500
Product Code. 11702964
Brand: BD 231342
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Oxytetracycline
Concentration 30 μg
Packaging Type Disc
Quantity 10 Pack