Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Novobiocin (30μg)
Product Code. 11762954
Click to view available options
Concentration:
30 μg
Unit Size:
Pack of 500
Product Code. 11762954
Brand: BD 231315
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Novobiocin
Concentration 30 μg
Packaging Type Disc
Quantity 10 Pack
compliance-icons
Product Identifier
  • BD BBL Sensi-Disc Novobiocin 30
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : sodium novobiocin