Learn More
BD BBL™ Sensi-Disc™ Elution Discs for Susceptibility Testing of Anaerobes
Click to view available options
Concentration:
80 μg
Unit Size:
Pack of 10
Specifications
Specifications
| Antimicrobial Agents | Metronidazole 78 |
| Concentration | 80 μg |
| Packaging Type | Disc |
| Quantity | 10 Pack |
For in vitro Diagnostic Use.