Learn More
BD BBL™ Sensi-Disc™ Elution Discs for Susceptibility Testing of Anaerobes
Product Code. 17302391
Click to view available options
Concentration:
80 μg
Unit Size:
Pack of 10
Product Code. 17302391
Brand: BD 231605
For determining the in vitro susceptibility of anaerobic bacteria to metronidazole by the broth-disc method
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Metronidazole 78
Concentration 80 μg
Packaging Type Disc
Quantity 10 Pack
For in vitro Diagnostic Use.