Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Ceftazidime23
Product Code. 17352391
Click to view available options
Quantity:
10 Pack
Unit Size:
Pack of 10
Product Code. 17352391
Brand: BD 231633
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Ceftazidime
Concentration 30 μg
Packaging Type Disc
Quantity 10 Pack
compliance-icons
Product Identifier
  • BD BBL Sensi-Disc Ceftazidime 30
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
Hazard Statement
  • H300-Fatal if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Ceftazidime
For in vitro diagnostic use