Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Disc, Ticarcillin w/Clavulanic Acid70
Click to view available options
Quantity:
10 Pack
Unit Size:
Pack of 10
Description
- 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
- Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Specifications
| Antimicrobial Agents | Clavulanic Acid70 |
| Concentration | 75/10 μg |
| Packaging Type | Disc |
| Quantity | 10 Pack |
For in vitro diagnostic use