Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Disc, Ticarcillin w/Clavulanic Acid70
Product Code. 17392391
Click to view available options
Quantity:
10 Pack
Unit Size:
Pack of 10
Product Code. 17392391
Brand: BD 231649
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Clavulanic Acid70
Concentration 75/10 μg
Packaging Type Disc
Quantity 10 Pack
For in vitro diagnostic use