Learn More
BD BBL™ Mycoflask™ Thioglycollate Prepared Media Enriched with Vitamin K1 and Hemin
Product Code. 17332351
Click to view available options
Quantity:
10 x 8 mL
100 x 5 mL
100 x 8 mL
Color:
Beige
Light Yellow
Packaging Type:
Size K tube with tight cap
Unit Size:
Pack of 10
Pack of 100
Product Code. 17332351
Brand: BD 221787
Ideal for use in the isolation and cultivation of fastidious or slow growing, obligately anaerobic microorganisms present in clinical materials
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE, BS12, CLSI, CMPH2, MCM9
Color Light Yellow
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Thioglycollate Medium
Formulation Pancreatic digest of Casein 17g/L, Papaic digest of Soybean Meal 3g/L, Dextrose 6g/L, Sodium Chloride 2.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.7g/L, L-Cystine 0.25g/L, Sodium Sulfite 0.1g/L, Hemin 0.005g/L, Vitamin K1 0.001g/L
For Use With (Application) Cultivation of fastidious and non-fastidious microorganisms
Packaging Type Size K tube with tight cap
Quantity 10 x 8 mL
For in vitro diagnostic use.