missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Haemophilus Test Prepared Plated Media Agar
Click to view available options
:
Haemophilus test media Agar; 8/Pk.
Description
- Ready-to-use
- 150 x 15mm-style plates
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Light Beige |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Haemophilus test media agar |
| Formulation | Beef extract 2g/L, Acid Hydrolysate of Casein 17.5g/L, Starch 1.5g/L, Agar 17g/L, Yeast extract 5%, Hematin 15mg/L Nicotinamide Adenine Dinucleotide 15mg/L |
| For Use With (Application) | Antimicrobial disc diffusion susceptibility procedure for Haemophilus species |
| Packaging Type | Plate |
| Quantity | 8/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction